Läs mer
25-Hydroxyvitamin D2 3-Hemisuccinate, TRC
Produktkod. 30134467
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134467 10 mg
30003532 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134467 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH99577010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1612177-56-6
Molekylformel C32H48O5
Molekylvikt (g/mol) 512.72
Synonym Butanedioic Acid 1-[(1S,3Z)-4-Methylene-3-[(2E)-2-[(1R,3aS,7aR)-octahydro-1-[(1R,2E,4S)-5-hydroxy-1,4,5-trimethyl-2-hexen-1-yl]-7a-methyl-4H-inden-4-ylidene]ethylidene]cyclohexyl] Ester, 25-OH Vitamin D2 3-Succinate
LEDER C[C@H](\C=C\[C@H](C)C(C)(C)O)[C@H]1CC[C@H]2\C(=C\C=C/3\C[C@H](CCC3=C)OC(=O)CCC(=O)O)\CCC[C@]12C
Specifikationer
Kemiskt namn eller material 25-Hydroxyvitamin D2 3-Hemisuccinate