Läs mer
Tocris Bioscience™ Pomalidomide 4'-alkylC5-acid
Kvantitet:
25 mg
Beskrivning
Pomalidomide 4'-alkylC5-acid is a functionalized cereblon ligand for PROTAC® research and development; incorporates an E3 ligase ligand plus an alkylC5 linker with terminal carboxylic acid ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
This product has been recently renamed. The previous name for this product was Pomalidomide - linker 3
Please contact us for SD files of our available Degrader Building Blocks.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifikationer
Specifikationer
| Kemiskt namn eller material | 6-[[2-(2,6-Dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1H-isoindol-4-yl]amino]hexanoic acid |
| CAS | 2225940-49-6 |
| Synonym | Pomalidomide - linker 3 |
| Kvantitet | 25 mg |
| Molekylformel | C19H21N3O6 |
| Formel vikt | 387.39 |
| Produkttyp | Ligand |
| Renhet | 95% |