Läs mer
25R-3Alpha,7Alpha,12Alpha-Trihydroxy-5Beta-cholestanoic Acid, TRC
Produktkod. 30454798
Change view
Kvantitet:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30454798 10 mg
30449859 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30454798 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT88796510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 23740-14-9
Molekylformel C27H46O5
Molekylvikt (g/mol) 450.65
Synonym (25R)-3α,7α,12α-Trihydroxy-5β-cholestan-26-oic Acid, (3α,5β,7α,12α,25R)-3,7,12-Trihydroxy-5β-cholestan-26-oic Acid
IUPAC-namn (2R,6R)-2-methyl-6-[(3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]heptanoic acid
LEDER O[C@@H]1CC[C@@]2(C)[C@@](C[C@@H](O)[C@]3([H])[C@]2([H])C[C@H](O)[C@@]4(C)[C@@]3([H])CC[C@]4([H])[C@H](C)CCC[C@@H](C)C(O)=O)([H])C1
Specifikationer
Kemiskt namn eller material 25R-3alpha,7alpha,12alpha-Trihydroxy-5beta-cholestanoic Acid
Kvantitet 10 mg
InChI formel InChI=1S/C27H46O5/c1-15(6-5-7-16(2)25(31)32)19-8-9-20-24-21(14-23(30)27(19,20)4)26(3)11-10-18(28)12-17(26)13-22(24)29/h15-24,28-30H,5-14H2,1-4H3,(H,31,32)/t15-,16-,17+,18-,19-,20+,21+,22-,23+,24+,26+,27-/m1/s1
Formel vikt 450.3345