missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Digoxigenin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB75201-SP
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Digoxigenin Monoclonal specifically detects Digoxigenin in Multi-Species samples. It is validated for Western Blot.
Specifikationer
| Digoxigenin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Detects Digoxigenin labeled proteins, nucleic acids and Digoxigenin conjugated primary and secondary antibodies. | |
| Multi-species | |
| Purified |
| Western Blot | |
| 611532 | |
| Western Blot 0.1 ug/mL | |
| DIG | |
| KLH-coupled Digoxigenin | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering