missing translation for 'onlineSavingsMsg'
Läs mer

FAT1 Antibody (FAT-1 3D7/1), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NB100-2693-0.025mg

 Se fler versioner av denna produkt

Produktkod. 18483299

  • 1825.00 SEK / 25µg

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Beskrivning

Beskrivning

FAT1 Monoclonal specifically detects FAT1 in Drosophila samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifikationer

Specifikationer

FAT1
Monoclonal
1 mg/mL
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500
FAT1
Cytoplasmic domain of drosophila Fat
0.025 mg
Signal Transduction
2195
Drosophila
Purified
Western Blot
FAT-1 3D7/1
Unconjugated
FAT tumor suppressor 1, FAT tumor suppressor homolog 1 (Drosophila)
Mouse
Thiophilic Chromatography
RUO
Primary
FAT (Fat-1). Tested only in Drosophila. Expected to cross-react with Rat (85% identity with immunogen) and Chimpanzee (92% identity with immunogen) due to sequence homology.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
IgM
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only