missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Human/Mouse Crk Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4316-SP
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Crk Monoclonal specifically detects Crk in Human, Mouse samples. It is validated for Western Blot.
Specifikationer
| Crk | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P46108 | |
| CRK | |
| E. coli-derived recombinant human Crk Gln205-Ser304 Accession # P46108 | |
| 25 μg | |
| Primary | |
| Detects human Crk in direct ELISAs and human and mouse Crk in Western blots. | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| 463414 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| adapter molecule crk, avian sarcoma virus CT10 (v-crk) oncogene homolog, CRKII, p38, Proto-oncogene c-Crk, v-crk avian sarcoma virus CT10 oncogene homolog, v-crk sarcoma virus CT10 oncogene homolog (avian) | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1398 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering