Learn More
27-Oxo-fusidic Acid, TRC
Product Code. 30415150
Change view
Click to view available options
Kvantitet:
10 mg
Product Code. Kvantitet
30415150 10 mg
1 options
Product Code. 30415150 Supplier Toronto Research Chemicals Supplier No. TRCO85685510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material 26-Oxofusidic Acid
CAS 1415035-94-7
Molekylformel C31 H46 O7
Kvantitet 10 mg
Synonym 29-Nordammara-17(20),24-dien-21-oic acid','16-(acetyloxy)-3,11-dihydroxy-26-oxo-','(3α,4α,8α,9β,11α,13α,14β,16β,17Z)-, ent-(17Z,24EZ)-16α-(Acetyloxy)-3β,11β-dihydroxy-4β,8,14-trimethyl-26-oxo-18-nor-5β,10α-cholesta-17(20),24-dien-21-oic acid, 26-Oxofusidic acid, 27-Oxo-fusidic acid
InChI formel InChI=1S/C31H46O7/c1-17(16-32)8-7-9-20(28(36)37)26-22-14-24(35)27-29(4)12-11-23(34)18(2)21(29)10-13-30(27,5)31(22,6)15-25(26)38-19(3)33/h8,16,18,21-25,27,34-35H,7,9-15H2,1-6H3,(H,36,37)/b17-8?,26-20-/t18-,21-,22-,23+,24+,25-,27-,29-,30-,31-/m0/s1
LEDER C[C@@H]1[C@H](O)CC[C@@]2(C)[C@H]1CC[C@@]3(C)[C@H]2[C@H](O)C[C@H]4\C(=C(/CCC=C(C)C=O)\C(=O)O)\[C@H](C[C@]34C)OC(=O)C
IUPAC-namn (2Z)-2-[(3R,4S,5S,8S,9S,10S,11R,13R,14S,16S)-16-acetyloxy-3,11-dihydroxy-4,8,10,14-tetramethyl-2,3,4,5,6,7,9,11,12,13,15,16-dodecahydro-1H-cyclopenta[a]phenanthren-17-ylidene]-6-methyl-7-oxohept-5-enoic acid
Molekylvikt (g/mol) 530.6927
Formel vikt 530.3244