missing translation for 'onlineSavingsMsg'
Läs mer

2alpha-Hydroxymethyl Ethisterone, TRC

Produktkod. 30009269
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30009269 25 mg
30009313 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30009269 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCH94669525MG

Please to purchase this item. Need a web account? Register with us today!

Denna vara kräver en prisuppdatering innan den beställs.

Begär ett pris för denna vara här.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2787-03-3
Molekylformel C22H30O3
Synonym (2α,17α)-17-Hydroxy-2-(hydroxymethyl)pregn-4-en-20-yn-3-one;
IUPAC-namn (2S,8R,9S,10R,13S,14S,17R)-17-ethynyl-17-hydroxy-2-(hydroxymethyl)-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one
LEDER C[C@]12C[C@@H](CO)C(=O)C=C1CC[C@@H]3[C@@H]2CC[C@@]4(C)[C@H]3CC[C@@]4(O)C#C

Specifications

Kemiskt namn eller material 2alpha-Hydroxymethyl Ethisterone
Kvantitet 25 mg
InChI formel InChI=1S/C22H30O3/c1-4-22(25)10-8-18-16-6-5-15-11-19(24)14(13-23)12-20(15,2)17(16)7-9-21(18,22)3/h1,11,14,16-18,23,25H,5-10,12-13H2,2-3H3/t14-,16+,17-,18-,20-,21-,22-/m0/s1
Formel vikt 342.2195
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.