Läs mer
(2E)-1-(2,4,6-Trichlorophenyl)-2-propanone O-Methyloxime, TRC
Produktkod. 30452875
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30452875 25 mg
1 options
Produktkod. 30452875 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT77453825MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (2E)-1-(2,4,6-Trichlorophenyl)-2-propanone O-Methyloxime
CAS 1452295-74-7
Molekylformel C10H10Cl3NO
Kvantitet 25 mg
Synonym (E)-1-(2,4,6-Trichlorophenyl)propan-2-one O-Methyl Oxime
InChI formel InChI=1S/C10H10Cl3NO/c1-6(14-15-2)3-8-9(12)4-7(11)5-10(8)13/h4-5H,3H2,1-2H3/b14-6+
LEDER CO\N=C(/C)\Cc1c(Cl)cc(Cl)cc1Cl
IUPAC-namn (E)-N-methoxy-1-(2,4,6-trichlorophenyl)propan-2-imine
Molekylvikt (g/mol) 266.55
Formel vikt 264.9828