Läs mer
(2E)-3-Methyl-2-hexenoic Acid, TRC
Produktkod. 30482383
Change view
Kvantitet:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30482383 100 mg
30472615 50 mg
30471470 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30482383 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM336140100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 27960-21-0
Molekylformel C7H12O2
Molekylvikt (g/mol) 128.17
Synonym (E)-3-Methyl-2-hexenoic Acid, trans-3-Methyl-2-hexenoic Acid
IUPAC-namn (E)-3-methylhex-2-enoic acid
LEDER CCC\C(=C\C(=O)O)\C
Specifikationer
Kemiskt namn eller material (2E)-3-Methyl-2-hexenoic Acid
Kvantitet 100 mg
InChI formel InChI=1S/C7H12O2/c1-3-4-6(2)5-7(8)9/h5H,3-4H2,1-2H3,(H,8,9)/b6-5+
Formel vikt 128.0837