Läs mer
(2E,4E)-2,4-Tetradecadienoic acid, TRC
Produktkod. 30424055
Change view
Kvantitet:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30424055 50 mg
30418336 25 mg
30416107 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30424055 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT29240050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 24738-49-6
Molekylformel C14 H24 O2
Molekylvikt (g/mol) 224.34
Synonym (2E,4E)-2,4-Tetradecadienoic acid, (2E,4E)-Tetradecadienoic acid, (E,E)-Tetradeca-2,4-dienoic acid
IUPAC-namn (2E,4E)-tetradeca-2,4-dienoic acid
LEDER CCCCCCCCC\C=C\C=C\C(=O)O
Specifikationer
Kemiskt namn eller material (2E,4E)-2,4-Tetradecadienoic acid
Kvantitet 50 mg
InChI formel InChI=1S/C14H24O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14(15)16/h10-13H,2-9H2,1H3,(H,15,16)/b11-10+,13-12+
Formel vikt 224.1776