Läs mer
(2E,4E)-5-[(2S,3S)-3-[(3S)-3,4-Dihydroxy-4-methylpentyl]-3-methyl-2-oxiranyl]-3-methyl-2,4-pentadienoic Acid Methyl Ester, TRC
Produktkod. 30378965
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Kvantitet:
10 mg
Produktkod. Kvantitet
30378965 10 mg
1 missing translation for 'options'
Produktkod. 30378965 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCD45377510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (2E,4E)-5-[(2S,3S)-3-[(3S)-3,4-Dihydroxy-4-methylpentyl]-3-methyl-2-oxiranyl]-3-methyl-2,4-pentadienoic Acid Methyl Ester
CAS 153651-94-6
Molekylformel C14 H19 N3 O5 S
Kvantitet 10 mg
Synonym [2S-[2alpha(2E,4E),3beta(R*)]]-5-[3-(3,4-Dihydroxy-4-methylpentyl)-3-methyloxiranyl]-3-methyl-2,4-Pentadienoic acid
InChI formel InChI=1S/C14H19N3O5S/c1-6(18)9-12(20)17-10(14(21)22)8(23-13(9)17)5-16-4-2-3-7(16)11(15)19/h6-7,9,13,18H,2-5H2,1H3,(H2,15,19)(H,21,22)/t6-,7+,9+,13-/m1/s1
LEDER C[C@@H](O)[C@@H]1[C@H]2SC(=C(N2C1=O)C(=O)O)CN3CCC[C@H]3C(=O)N
IUPAC-namn (5R,6S)-3-[[(2S)-2-carbamoylpyrrolidin-1-yl]methyl]-6-[(1R)-1-hydroxyethyl]-7-oxo-4-thia-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid
Molekylvikt (g/mol) 341.383
Formel vikt 341.105