Läs mer
(2E,4E,6E)-2,4,6-Nonatrienal (>90%), TRC
Produktkod. 30470855
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30470855 1 g
30467702 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30470855 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN6495401G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 57018-53-8
Molekylformel C9 H12 O
Molekylvikt (g/mol) 136.19
Synonym (E,E,E)-2,4,6-Nonatrienal, (E,E,E)-Nona-2,4,6-trienal, trans,trans,trans-Nona-2,4,6-trienal
IUPAC-namn (2E,4E,6E)-nona-2,4,6-trienal
LEDER CC\C=C\C=C\C=C\C=O
Specifikationer
Kemiskt namn eller material (2E,4E,6E)-2,4,6-Nonatrienal
Kvantitet 1 g
InChI formel InChI=1S/C9H12O/c1-2-3-4-5-6-7-8-9-10/h3-9H,2H2,1H3/b4-3+,6-5+,8-7+
Formel vikt 136.0888