Läs mer
(2E)-N-(2-Methylphenyl)-2-butenamide, TRC
Product Code. 30465888
Change view
Click to view available options
Kvantitet:
10 g
Product Code. Kvantitet
30465888 10 g
1 options
Product Code. 30465888 Supplier Toronto Research Chemicals Supplier No. TRCM32687510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material (2E)-N-(2-Methylphenyl)-2-butenamide
CAS 2196187-17-2
Molekylformel C11H13NO
Kvantitet 10 g
Synonym 2-Butenamide, N-(2-methylphenyl)-, (2E)- (ACI), (2E)-N-(2-Methylphenyl)-2-butenamide (ACI), (2E)-N-(2-Methylphenyl)but-2-enamide, Crotamiton Imp.C (EP)
InChI formel InChI=1S/C11H13NO/c1-3-6-11(13)12-10-8-5-4-7-9(10)2/h3-8H,1-2H3,(H,12,13)/b6-3+
LEDER CC=CC(=O)Nc1ccccc1C
IUPAC-namn (E)-N-(2-methylphenyl)but-2-enamide
Molekylvikt (g/mol) 175.23
Formel vikt 175.0997