Läs mer
(2E)-Octadecenal (>90%), TRC
Produktkod. 30138587
Change view
Kvantitet:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138587 500 mg
30137607 250 mg
30012746 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138587 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO233700500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 51534-37-3
Molekylformel C18 H34 O
Molekylvikt (g/mol) 266.46
Synonym Koiganal I, trans-2-Octadecenal, (E)-2-Octadecenal;
IUPAC-namn (E)-octadec-2-enal
LEDER CCCCCCCCCCCCCCC\C=C\C=O
Specifikationer
Kemiskt namn eller material (2E)-Octadecenal
Kvantitet 500 mg
InChI formel InChI=1S/C18H34O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h16-18H,2-15H2,1H3/b17-16+
Formel vikt 266.261