Läs mer
2H-1,3-Benzodithiole-2-thione, TRC
Produktkod. 30330333
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30330333 250 mg
1 options
Produktkod. 30330333 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB346150250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2H-1,3-Benzodithiole-2-thione
CAS 934-36-1
Molekylformel C7H4S3
Kvantitet 250 mg
Synonym Benzoisotrithione, Cyclic o-Phenylene Trithiocarbonate Dithiocatechol Trithiocarbonate, o-Phenylene Trithiocarbonate
InChI formel InChI=1S/C7H4S3/c8-7-9-5-3-1-2-4-6(5)10-7/h1-4H
LEDER S=C1Sc2ccccc2S1
IUPAC-namn 1,3-benzodithiole-2-thione
Molekylvikt (g/mol) 184.3