Läs mer
2H-2-Ethyl Candesartan Cilexetil, TRC
Produktkod. 30136753
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136753 250 mg
18030950 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136753 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE900875250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 914613-36-8
Molekylformel C35 H38 N6 O6
Molekylvikt (g/mol) 638.71
Synonym 1H-Benzimidazole-7-carboxylic acid, 2-ethoxy-1-[[2'-(2-ethyl-2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-, 1-[[(cyclohexyloxy)carbonyl]oxy]ethyl ester, (1RS)-1-[[(Cyclohexyloxy)carbonyl]-oxy]ethyl 2-Ethoxy-1-[[2'-(2-ethyl-2H-tetrazol-5-yl)-biphenyl-4-yl]methyl]-1H-benzimidazole-7-carboxylate, Candesartan Cilexetil Imp. F (EP)
IUPAC-namn 1-cyclohexyloxycarbonyloxyethyl 2-ethoxy-3-[[4-[2-(2-ethyltetrazol-5-yl)phenyl]phenyl]methyl]benzimidazole-4-carboxylate
LEDER CCOc1nc2cccc(C(=O)OC(C)OC(=O)OC3CCCCC3)c2n1Cc4ccc(cc4)c5ccccc5c6nnn(CC)n6
Specifikationer
Kemiskt namn eller material 2H-2-Ethyl Candesartan Cilexetil
Anteckningar om renhetsgrad HPLC
Kvantitet 250 mg
InChI formel InChI=1S/C35H38N6O6/c1-4-41-38-32(37-39-41)28-15-10-9-14-27(28)25-20-18-24(19-21-25)22-40-31-29(16-11-17-30(31)36-34(40)44-5-2)33(42)45-23(3)46-35(43)47-26-12-7-6-8-13-26/h9-11,14-21,23,26H,4-8,12-13,22H2,1-3H3
Formel vikt 638.2853
Procent renhet >95%