Läs mer
2H,5H-Pyrano[4,3-b]pyranyl Mupirocin Methyl Ester, TRC
Produktkod. 30417720
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30417720 1 mg
1 options
Produktkod. 30417720 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP8405251MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2H,5H-Pyrano[4,3-b]pyranyl Mupirocin Methyl Ester
CAS 71087-95-1
Molekylformel C27H46O9
Kvantitet 1 mg
Synonym [2R-[2α(1S*,2S*),3β,4aα,7β(E),8α,8aβ]]-9-[[4-[hexahydro-3,8-dihydroxy-2-(2-hydroxy-1-methylpropyl)-2H,5H-pyrano[4,3-b]pyran-7-yl]-3-methyl-1-oxo-2-butenyl]oxy]nonanoic Acid Methyl Ester
InChI formel InChI=1S/C27H46O9/c1-17(14-24(31)34-12-10-8-6-5-7-9-11-23(30)33-4)13-22-25(32)27-20(16-35-22)15-21(29)26(36-27)18(2)19(3)28/h14,18-22,25-29,32H,5-13,15-16H2,1-4H3/b17-14+/t18-,19-,20-,21-,22-,25-,26+,27+/m0/s1
LEDER O[C@@H]1[C@@]2([H])[C@@](CO[C@H]1C/C(C)=C/C(OCCCCCCCCC(OC)=O)=O)([H])C[C@@H]([C@H](O2)[C@@H](C)[C@@H](O)C)O
IUPAC-namn methyl 9-(((E)-4-((2R,3S,4aS,7S,8S,8aR)-3,8-dihydroxy-2-((2S,3S)-3-hydroxybutan-2-yl)hexahydro-2H,5H-pyrano[4,3-b]pyran-7-yl)-3-methylbut-2-enoyl)oxy)nonanoate
Molekylvikt (g/mol) 514.65
Formel vikt 514.31