Läs mer
2H,5H-Pyrano[4,3-b]pyranyl Mupirocin Sodium Impurity, TRC
Produktkod. 30416478
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30416478 5 mg
1 options
Produktkod. 30416478 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP8405505MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 9-[((2E)-4-[(2R,3RS,4aS,7S,8S,8aR)-3,8-Dihydroxy-2-[(1S,2S)-2-hydroxy-1-methylpropyl]hexahydro-2H,5H-pyrano[4,3-b]pyran-7-yl]-3-methylbut-2-enoyl)oxy]nonanoic Acid Sodium Salt
CAS 116182-44-6
Molekylformel C26 H43 O9 . Na
Kvantitet 5 mg
Synonym Nonanoic acid','9-[[4-[hexahydro-3,8-dihydroxy-2-(2-hydroxy-1-methylpropyl)-2H,5H-pyrano[4,3-b]pyran-7-yl]-3-methyl-1-oxo-2-butenyl]oxy]-','monosodium salt','[2R-[2α(1S*,2S*),3β,4aα,7β(E),8α,8aβ]]- (9CI), 2H,5H-Pyrano[4,3-b]pyran','nonanoic acid deriv., 9-[[(2E)-4-[(2R,3RS,4aS,7S,8S,8aR)-3,8-dihydroxy-2-[(1S,2S)-2-hydroxy-1-methylpropyl]hexahydro-2H,5H-pyrano[4,3-b]pyran-7-yl]-3-methylbut-2-enoyl]oxy]nonanoic acid sodium salt, Mupirocin Calcium Imp. E (EP) sodium salt, 2H,5H-Pyrano[4,3-b]pyranyl Mupirocin sodium impurity
InChI formel InChI=1S/C26H44O9.Na/c1-16(13-23(31)33-11-9-7-5-4-6-8-10-22(29)30)12-21-24(32)26-19(15-34-21)14-20(28)25(35-26)17(2)18(3)27;/h13,17-21,24-28,32H,4-12,14-15H2,1-3H3,(H,29,30);/q;+1/p-1/b16-13+;/t17-,18-,19-,20-,21-,24-,25+,26+;/m0./s1
LEDER [Na+].C[C@H](O)[C@H](C)[C@H]1O[C@H]2[C@@H](O)[C@H](C\C(=C\C(=O)OCCCCCCCCC(=O)[O-])\C)OC[C@@H]2C[C@@H]1O
IUPAC-namn sodium;9-[(E)-4-[(2R,3S,4aS,7S,8S,8aR)-3,8-dihydroxy-2-[(2S,3S)-3-hydroxybutan-2-yl]-2,3,4,4a,5,7,8,8a-octahydropyrano[4,3-b]pyran-7-yl]-3-methylbut-2-enoyl]oxynonanoate
Molekylvikt (g/mol) 522.6
Formel vikt 522.2805