Läs mer
2H-Perfluoro-2-dodecenoic Acid, TRC
Produktkod. 30133796
Change view
Kvantitet:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133796 250 mg
17914325 25 mg
17983795 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133796 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP271640250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 70887-94-4
Molekylformel C12 H2 F20 O2
Molekylvikt (g/mol) 558.11
Synonym 2-Dodecenoic acid, 3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-eicosafluoro-, 3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-Eicosafluoro-2-dodecenoic acid, 2H-Perfluoro-2-dodecenoate, 3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-Icosafluorododec-2-enoicacid, 10:2 FTUCA, 2H-Perfluoro-2-dodecenoic Acid
IUPAC-namn (Z)-3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-icosafluorododec-2-enoic acid
LEDER OC(=O)\C=C(/F)\C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Specifikationer
Kemiskt namn eller material 2H-Perfluoro-2-dodecenoic acid
Anteckningar om renhetsgrad HPLC
Kvantitet 250 mg
InChI formel InChI=1S/C12H2F20O2/c13-2(1-3(33)34)4(14,15)5(16,17)6(18,19)7(20,21)8(22,23)9(24,25)10(26,27)11(28,29)12(30,31)32/h1H,(H,33,34)/b2-1-
Formel vikt 557.9735
Procent renhet >95%