Läs mer
2OH-BNPP1, MedChemExpress
Produktkod. 16483708
Encompass_Preferred
Change view
Kvantitet:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16483708 10 mg
16476447 5 mg
16472836 25 mg
16450097 50 mg
16424786 100 mg
16420076 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Produktkod. 16483708 Leverantör Medchem Express Leverantörsnummer HY102081
MedChemExpress 2OH-BNPP1 is an inhibitor of BUB1 kinase, a Ser/Thr kinase, used for the treatment of cancer.
2OH-BNPP1 is an inhibitor of BUB1 kinase, a Ser/Thr kinase, used for the treatment of cancer.
Specifikationer
Kemiskt namn eller material 2OH-BNPP1
CAS 833481-73-5
Färg Off-White
Anteckningar om renhetsgrad Research
Molekylformel C16H19N5O
Kvantitet 10 mg
Löslighetsinformation DMSO : ≥ 100 mg/mL (336.30 mM)
LEDER OC1=CC=CC=C1CC2=NN(C(C)(C)C)C3=NC=NC(N)=C32
Molekylvikt (g/mol) 297.35
Formel vikt 297.35
Procent renhet 99.65%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-Kinase/protease