Läs mer
(2R)-2-[6-(4-Chlorophenoxy)hexyl]-oxiranecarboxylic Acid Sodium Salt, TRC
Produktkod. 30350461
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30350461 10 mg
30360759 25 mg
30366281 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30350461 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC43308010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 828934-41-4
Molekylformel C15 H18 Cl O4 . Na
Molekylvikt (g/mol) 320.744
IUPAC-namn sodium;(2R)-2-[6-(4-chlorophenoxy)hexyl]oxirane-2-carboxylate
LEDER [Na+].[O-]C(=O)[C@@]1(CCCCCCOc2ccc(Cl)cc2)CO1
Specifikationer
Kemiskt namn eller material (2R)-2-[6-(4-Chlorophenoxy)hexyl]-oxiranecarboxylic Acid Sodium Salt
Kvantitet 10 mg
InChI formel InChI=1S/C15H19ClO4.Na/c16-12-5-7-13(8-6-12)19-10-4-2-1-3-9-15(11-20-15)14(17)18;/h5-8H,1-4,9-11H2,(H,17,18);/q;+1/p-1/t15-;/m1./s1
Formel vikt 320.079