Läs mer
(2R)-2-Amino-2-(4-hydroxyphenyl)acetamide, Mikromol™
Produktkod. 30049519
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30049519 25 mg
30049164 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30049519 Leverantör Mikromol Leverantörsnummer MM0232.120025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2R)-2-Amino-2-(4-hydroxyphenyl)acetamide
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (2R)-2-Amino-2-(4-hydroxyphenyl)acetamide
CAS 54397-23-8
Analyt- eller komponentnamn (2R)-2-Amino-2-(4-hydroxyphenyl)acetamide
Frakt skick Room Temperature
Molekylformel C8 H10 N2 O2
Certifikat och efterlevnad ISO 17025
Synonym (2R)-2-Amino-2-(4-hydroxyphenyl)acetamide
InChI formel InChI=1S/C8H10N2O2/c9-7(8(10)12)5-1-3-6(11)4-2-5/h1-4,7,11H,9H2,(H2,10,12)/t7-/m1/s1
LEDER N[C@@H](C(=O)N)c1ccc(O)cc1
IUPAC-namn (2R)-2-amino-2-(4-hydroxyphenyl)acetamide
Molekylvikt (g/mol) 166.18
Kvantitet 25 mg
Formel vikt 166.0742 g/mol