Läs mer
(2R)-2-Hydroxyglutaric Acid Octyl Ester Sodium Salt, TRC
Produktkod. 17939254
Change view
Kvantitet:
1 mg
5 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17939254 50 mg
17949074 1 mg
17949994 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17939254 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH94259550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1391068-16-8
Molekylformel C13H23NaO5
Molekylvikt (g/mol) 282.31
Synonym (2R)-2-Hydroxypentanedioic Acid Octyl Ester Sodium Salt, (R)-2-Hydroxyglutaric Acid Octyl Ester Sodium Salt, (R)-α-Hydroxyglutaric Acid Octyl Ester Sodium Salt, D-2-Hydroxyglutaric Acid Octyl Ester Sodium Salt, Octyl D-2-Hydroxyglutarate Sodium Salt, Octyl-D-2-HG Sodium;
LEDER [Na+].CCCCCCCCOC(=O)[C@H](O)CCC(=O)[O-]
Specifikationer
Kemiskt namn eller material (2R)-2-Hydroxyglutaric Acid Octyl Ester Sodium Salt
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C13H24O5.Na/c1-2-3-4-5-6-7-10-18-13(17)11(14)8-9-12(15)16;/h11,14H,2-10H2,1H3,(H,15,16);/q;+1/p-1/t11-;/m1./s1
Formel vikt 282.1443
Procent renhet >95%