Läs mer
(2R)-2-Methyl-2-(phenylmethoxy)propanedioic Acid 1-Ethyl Ester, TRC
Produktkod. 30466778
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30466778 250 mg
1 options
Produktkod. 30466778 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM329265250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (2R)-2-Methyl-2-(phenylmethoxy)propanedioic Acid 1-Ethyl Ester
CAS 1786-91-0
Molekylformel C13H16O5
Kvantitet 250 mg
Synonym (R)-Methyl(phenylmethoxy)propanedioic Acid Monoethyl Ester
InChI formel InChI=1S/C13H16O5/c1-3-17-12(16)13(2,11(14)15)18-9-10-7-5-4-6-8-10/h4-8H,3,9H2,1-2H3,(H,14,15)/t13-/m1/s1
LEDER CCOC([C@](C(O)=O)(C)OCC1=CC=CC=C1)=O
IUPAC-namn (R)-2-(benzyloxy)-3-ethoxy-2-methyl-3-oxopropanoic acid
Molekylvikt (g/mol) 252.26
Formel vikt 252.1