Läs mer
(2R,3aR,6aR)-Ramipril, TRC
Produktkod. 30134734
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134734 50 mg
30011509 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134734 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR11103550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 129939-63-5
Molekylformel C23H32N2O5
Molekylvikt (g/mol) 416.51
Synonym (2R,3aR,6aR)-1-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-1-oxopropyl]octahydrocyclopenta[b]pyrrole-2-carboxylic Acid, [2R-[1[S*(S*)],2alpha,3abeta,6abeta]]-1-[2-[[1-(ethoxycarbonyl)-3-phenylpropyl]amino]-1-oxopropyl]octahydrocyclopenta[b]pyrrole-2-carboxylic Acid, Ramipril EP Impurity N;
IUPAC-namn (2R,3aR,6aR)-1-[(2S)-2-[[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino]propanoyl]-3,3a,4,5,6,6a-hexahydro-2H-cyclopenta[b]pyrrole-2-carboxylic acid
LEDER CCOC(=O)[C@H](CCc1ccccc1)N[C@@H](C)C(=O)N2[C@@H]3CCC[C@@H]3C[C@@H]2C(=O)O
Specifikationer
Kemiskt namn eller material (2R,3aR,6aR)-Ramipril
Kvantitet 50 mg
InChI formel InChI=1S/C23H32N2O5/c1-3-30-23(29)18(13-12-16-8-5-4-6-9-16)24-15(2)21(26)25-19-11-7-10-17(19)14-20(25)22(27)28/h4-6,8-9,15,17-20,24H,3,7,10-14H2,1-2H3,(H,27,28)/t15-,17+,18-,19+,20+/m0/s1
Formel vikt 416.2311