Läs mer
(2R,3S)-3-Amino-2-hydroxy-3-phenylpropanoic Acid Hydrochloride, Mikromol™
Produktkod. 30462538
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30462538 4 x 25 mg
30050883 100 mg
30049169 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30462538 Leverantör Mikromol Leverantörsnummer MM1489.0300254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2R,3S)-3-Amino-2-hydroxy-3-phenylpropanoic Acid Hydrochloride, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (2R,3S)-3-Amino-2-hydroxy-3-phenylpropanoic Acid Hydrochloride
CAS 132201-32-2
Analyt- eller komponentnamn (2R,3S)-3-Amino-2-hydroxy-3-phenylpropanoic Acid Hydrochloride
Frakt skick Room Temperature
Molekylformel C9 H11 N O3 . Cl H
Certifikat och efterlevnad ISO 17025
Synonym (2R,3S)-3-Phenylisoserine hydrochloride, Benzenepropanoic acid, beta-amino-alpha-hydroxy-, hydrochloride (1:1), (alphaR,betaS)-, Benzenepropanoic acid, beta-amino-alpha-hydroxy-, hydrochloride, (alphaR,betaS)- (9CI), Benzenepropanoic acid, beta-amino-alpha-hydroxy-, hydrochloride, [R-(R*,S*)]-
InChI formel InChI=1S/C9H11NO3.ClH/c10-7(8(11)9(12)13)6-4-2-1-3-5-6;/h1-5,7-8,11H,10H2,(H,12,13);1H/t7-,8+;/m0./s1
Lösningstyp Neat
LEDER Cl.N[C@H]([C@@H](O)C(=O)O)c1ccccc1
IUPAC-namn (2R,3S)-3-amino-2-hydroxy-3-phenylpropanoic acid;hydrochloride
Molekylvikt (g/mol) 217.65
Kvantitet 4 x 25 mg
Formel vikt 217.0506 g/mol