Läs mer
(2R,3S)-3-Phenylisoserine Ethyl Ester, TRC
Produktkod. 30410564
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30410564 1 g
1 options
Produktkod. 30410564 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP3354401G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (2R,3S)-3-Phenylisoserine Ethyl Ester
CAS 143615-00-3
Molekylformel C11H15NO3
Kvantitet 1 g
Synonym (2R,3S)-3-Amino-2-hydroxy-3-phenylpropionic Acid Ethyl Ester, (alphaR,betaS)-beta-Amino-alpha-hydroxy-benzenepropanoic Acid Ethyl Ester
InChI formel InChI=1S/C11H15NO3/c1-2-15-11(14)10(13)9(12)8-6-4-3-5-7-8/h3-7,9-10,13H,2,12H2,1H3/t9-,10+/m0/s1
LEDER CCOC(=O)[C@H](O)[C@@H](N)c1ccccc1
IUPAC-namn ethyl (2R,3S)-3-amino-2-hydroxy-3-phenylpropanoate
Molekylvikt (g/mol) 209.24
Formel vikt 209.1052