Learn More
(2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(octylthio)tetrahydro-2H-pyran-3,4,5-triol, 98%, BLD Pharm™
Spezifikation
Spezifikation
| CAS | 85618-21-9 |
| Molecular Formula | C14H28O5S |
| MDL Number | MFCD00012189 |
| Quantity | 5 g |
| SMILES | O[C@H]([C@H]([C@@H]([C@@H](CO)O1)O)O)[C@@H]1SCCCCCCCC |
| Molecular Weight (g/mol) | 308.43 |
| Percent Purity | 98% |
| Chemical Name or Material | (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(octylthio)tetrahydro-2H-pyran-3,4,5-triol |