Läs mer
(2R,4R)-1-[(2S)-2-[(tert-Butyloxycarbonyl)amino]-5-[[imino(nitroamino)methyl]amino]-1-oxopentyl]-4-methyl-2-piperidinecarboxylic Acid Benzyl Ester, TRC
Produktkod. 30344272
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30344272 10 mg
1 options
Produktkod. 30344272 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB69366510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (2R,4R)-1-((2S)-2-((tert-Butyloxycarbonyl)amino)-5-((imino(nitroamino)methyl)amino)-1-oxopentyl)-4-methyl-2-piperidinecarboxylic Acid Benzyl Ester
CAS 367952-82-7
Molekylformel C20 H25 N O3
Kvantitet 10 mg
Synonym (2R,4R)-1-[(2S)-2-[[(1,1-Dimethylethoxy)carbonyl]amino]-5-[[imino(nitroamino)methyl]amino]-1-oxopentyl]-4-methyl-2-piperidinecarboxylic Acid Phenylmethyl Ester
InChI formel InChI=1S/C20H25NO3/c21-18-11-19(24-13-16-9-5-2-6-10-16)17(20(18)22)14-23-12-15-7-3-1-4-8-15/h1-10,17-20,22H,11-14,21H2/t17-,18+,19+,20+/m1/s1
LEDER N[C@H]1C[C@H](OCc2ccccc2)[C@@H](COCc3ccccc3)[C@@H]1O
IUPAC-namn benzyl (2R,4R)-1-[(2S)-5-[[amino(nitramido)methylidene]amino]-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoyl]-4-methylpiperidine-2-carboxylate
Molekylvikt (g/mol) 327.417
Formel vikt 327.183