Läs mer
(2R,4R)-Sacubitril, TRC
Produktkod. 30006117
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30006117 1 mg
30003752 5 mg
30003723 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30006117 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS0809101MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 766480-48-2
Molekylformel C24 H29 N O5
Molekylvikt (g/mol) 411.491
Synonym (R*,R*)-γ-[(3-Carboxy-1-oxopropyl)amino]-α-methyl-[1,1’-biphenyl]-4-pentanoic Acid α-Ethyl Ester, (αR,γR)-rel-γ-[(3-Carboxy-1-oxopropyl)amino]-α-methyl-[1,1’-biphenyl]-4-pentanoic Acid 4-Ethyl Ester, Sacubitril-(2R,4R) Isomer;Impurity B
IUPAC-namn 4-[[(2R,4R)-5-ethoxy-4-methyl-5-oxo-1-(4-phenylphenyl)pentan-2-yl]amino]-4-oxobutanoic acid
LEDER CCOC(=O)[C@H](C)C[C@H](Cc1ccc(cc1)c2ccccc2)NC(=O)CCC(=O)O
Specifikationer
Kemiskt namn eller material (2R,4R)-Sacubitril
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C24H29NO5/c1-3-30-24(29)17(2)15-21(25-22(26)13-14-23(27)28)16-18-9-11-20(12-10-18)19-7-5-4-6-8-19/h4-12,17,21H,3,13-16H2,1-2H3,(H,25,26)(H,27,28)/t17-,21-/m1/s1
Formel vikt 411.205
Procent renhet >95%