missing translation for 'onlineSavingsMsg'
Learn More

(2R,5R)-1-[2-(1,3-Dioxolan-2-yl)phenyl]-2,5-diethylphospholane, 97%, Alfa Aesar™

Brand:  Alfa Aesar H63024.ME

Additional Details : CAS Number : 1217655-83-8 Weight : 0.00050kg

Quantity 500mg

 View more versions of this product

Product Code. 15498096

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Specifications

Chemical Identifiers

1217655-83-8
292.359
UKBFKBMQNUOKTD-KBPBESRZSA-N
16218309
CCC1CCC(P1C2=CC=CC=C2C3OCCO3)CC
C17H25O2P
MFCD09265098
2s,5s---1-2-1,3-dioxolan-2-yl phenyl-2,5-diethylphospholane, s,s-et-rajphos, 2-2-2s,5s-2,5-diethyl-1-phospholano phenyl 1,3-dioxolane, 2-2-2s,5s-2,5-diethylphospholan-1-yl phenyl-1,3-dioxolane, trans-1-2-1,3-dioxolan-2-yl phenyl-2,5-diethylphospholane, 2s,5s-1-2-1,3-dioxolan-2-yl phenyl-2,5-diethylphospholane, 2s,5s---1-2-1,3-dioxolan-2-yl phenyl-2,5-diethylphospholane, kanata purity
2-[2-[(2S,5S)-2,5-diethylphospholan-1-yl]phenyl]-1,3-dioxolane

Specifications

(2R,5R)-1-[2-(1,3-Dioxolan-2-yl)phenyl]-2,5-diethylphospholane
1.5620
Colorless to Yellow
1217655-83-8
MFCD09265098
UKBFKBMQNUOKTD-KBPBESRZSA-N
2-[2-[(2S,5S)-2,5-diethylphospholan-1-yl]phenyl]-1,3-dioxolane
16218309
Liquid
1.073g/mL
Air Sensitive
500mg
C17H25O2P
2s,5s---1-2-1,3-dioxolan-2-yl phenyl-2,5-diethylphospholane, s,s-et-rajphos, 2-2-2s,5s-2,5-diethyl-1-phospholano phenyl 1,3-dioxolane, 2-2-2s,5s-2,5-diethylphospholan-1-yl phenyl-1,3-dioxolane, trans-1-2-1,3-dioxolan-2-yl phenyl-2,5-diethylphospholane, 2s,5s-1-2-1,3-dioxolan-2-yl phenyl-2,5-diethylphospholane, 2s,5s---1-2-1,3-dioxolan-2-yl phenyl-2,5-diethylphospholane, kanata purity
CCC1CCC(P1C2=CC=CC=C2C3OCCO3)CC
292.359
292.35
97%
Videos
SDS Documents