missing translation for 'onlineSavingsMsg'
Learn More

(2R,5S)-L-Menthol-5-(acetyloxy)-1,3-oxathiolane-2-carboxylate, TRC

Product Code. 30472340
Change view
Click to view available options
Kvantitet:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30472340 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30472340 Supplier Toronto Research Chemicals Supplier No. TRCM21901110MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material (2R,5S)-L-Menthol-5-(acetyloxy)-1,3-oxathiolane-2-carboxylate
CAS 147126-65-6
Molekylformel C16 H26 O5 S
Kvantitet 10 mg
Synonym 1,3-Oxathiolane-2-carboxylic acid, 5-(acetyloxy)-, (1R,2S,5R)-5-methyl-2-(1-methylethyl)cyclohexyl ester, (2R,5S)-, 1,3-Oxathiolane-2-carboxylic acid, 5-(acetyloxy)-, 5-methyl-2-(1-methylethyl)cyclohexyl ester, [1R-[1α(2R*,5S*),2β,5α]]-, (2R,5S)-L-Menthol-5-(acetyloxy)-1,3-oxathiolane-2-carboxylate, (2R-cis)-5-Acetoxy-1,3-oxathiolane-2-carboxylic Acid (1R,2S,5R)-Menthyl Ester
InChI formel InChI=1S/C16H26O5S/c1-9(2)12-6-5-10(3)7-13(12)20-15(18)16-21-14(8-22-16)19-11(4)17/h9-10,12-14,16H,5-8H2,1-4H3/t10-,12+,13-,14+,16-/m1/s1
LEDER CC(C)[C@@H]1CC[C@@H](C)C[C@H]1OC(=O)[C@@H]2O[C@@H](CS2)OC(=O)C
IUPAC-namn [(1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexyl] (2R,5S)-5-acetyloxy-1,3-oxathiolane-2-carboxylate
Molekylvikt (g/mol) 330.44
Formel vikt 330.1501
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.