missing translation for 'onlineSavingsMsg'
Läs mer

Tocris Bioscience™ 2R,6R-Hydroxynorketamine hydrochloride

Produktkod. 18045082
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18045082 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18045082 Leverantör Tocris Bioscience™ Leverantörsnummer 6094/10

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Enhances AMPA currents; decreases D-serine (a NMDA co-agonist); lacks ketamine-related side effects

2R,6R-Hydroxynorketamine hydrochloride is a ketamine metabolite. It enhances AMPA receptor-mediated excitatory post-synaptic potentials in the CA1 region of hippocampal slices and decreases intracellular DL>D-serine (a NMDA co-agonist) concentrations in PC-12 cells (IC50 = 0.68 nM). 2R,6R-Hydroxynorketamine upregulates KCNQ2 channels in mouse ventral hippocampal glutamatergic neurons in vitro and in vivo. Exerts antidepressant effects in mice. Lacks ketamine-related side effects. 2S,6S-Enantiomer and Racemate also available.
TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material (2R,6R)-2-Amino-2-(2-chlorophenyl)-6-hydroxycyclohexanone hydrochloride
CAS 1430202-69-9
Kvantitet 10 mg
Mål AMPA Receptor Agonists
Molekylformel C12H14ClNO2.HCl
Renhet 98%
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.