Läs mer
(2RS)-1-[2-(2-Methoxyethyl)phenoxy]-3-[(1-methylethyl)amino]propan-2-ol, Mikromol™
Produktkod. 30460669
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30460669 4 x 25 mg
30050662 25 mg
30051250 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30460669 Leverantör Mikromol Leverantörsnummer MM0027.0700254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2RS)-1-[2-(2-Methoxyethyl)phenoxy]-3-[(1-methylethyl)amino]propan-2-ol, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (2RS)-1-[2-(2-Methoxyethyl)phenoxy]-3-[(1-methylethyl)amino]propan-2-ol
CAS 163685-38-9
Analyt- eller komponentnamn (2RS)-1-[2-(2-Methoxyethyl)phenoxy]-3-[(1-methylethyl)amino]propan-2-ol
Frakt skick Room Temperature
Molekylformel C15 H25 N O3
Certifikat och efterlevnad ISO 17025
Synonym Metoprolol Succinate Imp. E (EP), Metoprolol Tartrate Imp. E (EP), Metoprolol Imp. E (EP), (2RS)-1-[2-(2-Methoxyethyl)phenoxy]-3-[(1-methylethyl)amino]propan-2-ol, Metoprolol Succinate Impurity E, Metoprolol Tartrate Impurity E, Metoprolol Impurity E, Metoprolol Impurity E
InChI formel InChI=1S/C15H25NO3/c1-12(2)16-10-14(17)11-19-15-7-5-4-6-13(15)8-9-18-3/h4-7,12,14,16-17H,8-11H2,1-3H3
Lösningstyp Neat
LEDER COCCc1ccccc1OCC(O)CNC(C)C
IUPAC-namn 1-[2-(2-methoxyethyl)phenoxy]-3-(propan-2-ylamino)propan-2-ol
Molekylvikt (g/mol) 267.36
Kvantitet 4 x 25 mg
Formel vikt 267.1834 g/mol