Läs mer
(2RS)-1-(Isopropylamino)-3-[4-(2-methoxyethoxy)methylphenoxy]propan-2-ol, Mikromol™
Produktkod. 30460890
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30460890 4 x 25 mg
17941256 100 mg
17993856 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30460890 Leverantör Mikromol Leverantörsnummer MM0460.0500254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2RS)-1-(Isopropylamino)-3-[4-(2-methoxyethoxy)methylphenoxy]propan-2-ol, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (2RS)-1-(Isopropylamino)-3-[4-(2-methoxyethoxy)methylphenoxy]propan-2-ol
CAS 1346604-00-9
Analyt- eller komponentnamn (2RS)-1-(Isopropylamino)-3-[4-(2-methoxyethoxy)methylphenoxy]propan-2-ol
Frakt skick Room Temperature
Molekylformel C16 H27 N O4
Certifikat och efterlevnad ISO 17025
Synonym Bisoprolol Fumarate Imp. Q (EP), Bisoprolol Imp. Q (EP), (2RS)-1-(Isopropylamino)-3-[4-(2-methoxyethoxy)methylphenoxy]propan-2-ol, Bisoprolol Fumarate Impurity Q, Bisoprolol Impurity Q
InChI formel InChI=1S/C9H11FN2O5/c1-3-5(13)6(14)8(17-3)12-2-4(10)7(15)11-9(12)16/h2-3,5-6,8,13-14H,1H3,(H,11,15,16)/t3-,5-,6-,8-/m1/s5
Lösningstyp Neat
LEDER COCCOCc1ccc(OCC(O)CNC(C)C)cc1
IUPAC-namn 1-[4-(2-methoxyethoxymethyl)phenoxy]-3-(propan-2-ylamino)propan-2-ol
Molekylvikt (g/mol) 297.39
Kvantitet 4 x 25 mg
Formel vikt 297.194 g/mol