Läs mer
(2RS)-2-(2,4-Difluorophenyl)-1-(1H-1,2,4-triazol-1-yl)-3-(4H-1,2,4-triazol-4-yl)propan-2-ol, Mikromol™
Produktkod. 30458707
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30458707 4 x 25 mg
17971826 25 mg
17981116 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30458707 Leverantör Mikromol Leverantörsnummer MM0235.1100254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2RS)-2-(2,4-Difluorophenyl)-1-(1H-1,2,4-triazol-1-yl)-3-(4H-1,2,4-triazol-4-yl)propan-2-ol, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (2RS)-2-(2,4-Difluorophenyl)-1-(1H-1,2,4-triazol-1-yl)-3-(4H-1,2,4-triazol-4-yl)propan-2-ol
CAS 89429-59-4
Analyt- eller komponentnamn (2RS)-2-(2,4-Difluorophenyl)-1-(1H-1,2,4-triazol-1-yl)-3-(4H-1,2,4-triazol-4-yl)propan-2-ol
Frakt skick Room Temperature
Molekylformel C13 H12 F2 N6 O
Certifikat och efterlevnad ISO 17025
Synonym Fluconazole Imp. A (EP), (2RS)-2-(2,4-Difluorophenyl)-1-(1H-1,2,4-triazol-1-yl)-3-(4H-1,2,4-triazol-4-yl)propan-2-ol, Fluconazole Impurity A
InChI formel InChI=1S/C13H12F2N6O/c14-10-1-2-11(12(15)3-10)13(22,4-20-8-17-18-9-20)5-21-7-16-6-19-21/h1-3,6-9,22H,4-5H2
Lösningstyp Neat
LEDER OC(Cn1cnnc1)(Cn2cncn2)c3ccc(F)cc3F
IUPAC-namn 2-(2,4-difluorophenyl)-1-(1,2,4-triazol-1-yl)-3-(1,2,4-triazol-4-yl)propan-2-ol
Molekylvikt (g/mol) 306.27
Kvantitet 4 x 25 mg
Formel vikt 306.1041 g/mol