Läs mer
(2RS)-2-(2-Oxopyrrolidin-1-yl)butanoic Acid, Mikromol™
Produktkod. 30050565
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30050565 25 mg
30050329 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30050565 Leverantör Mikromol Leverantörsnummer MM1372.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2RS)-2-(2-Oxopyrrolidin-1-yl)butanoic Acid
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-(2-oxopyrrolidin-1-yl)butanoic acid
CAS 67118-31-4
Analyt- eller komponentnamn 2-(2-oxopyrrolidin-1-yl)butanoic acid
Frakt skick Room Temperature
Molekylformel C8 H13 N O3
Certifikat och efterlevnad ISO 17025
Synonym Levetiracetam Imp. A (EP), 1-Pyrrolidineacetic acid, alpha-ethyl-2-oxo-, Levetiracetam Impurity A
InChI formel InChI=1S/C8H13NO3/c1-2-6(8(11)12)9-5-3-4-7(9)10/h6H,2-5H2,1H3,(H,11,12)
LEDER CCC(N1CCCC1=O)C(=O)O
IUPAC-namn 2-(2-oxopyrrolidin-1-yl)butanoic acid
Molekylvikt (g/mol) 171.19
Kvantitet 25 mg
Formel vikt 171.0895 g/mol