Läs mer
(2RS)-2-(Dimethylamino)-2-phenylbutanol Hydrochloride, Mikromol™
Produktkod. 17974296
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17974296 25 mg
17902346 100 mg
30463178 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17974296 Leverantör Mikromol Leverantörsnummer MM0417.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2RS)-2-(Dimethylamino)-2-phenylbutanol Hydrochloride
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (2RS)-2-(Dimethylamino)-2-phenylbutanol Hydrochloride
CAS 60577-23-3
Analyt- eller komponentnamn (2RS)-2-(Dimethylamino)-2-phenylbutanol Hydrochloride
Frakt skick Room Temperature
Molekylformel C12 H19 N O . Cl H
Certifikat och efterlevnad ISO 17025
Synonym Trimebutine Imp. A (EP), (2RS)-2-(Dimethylamino)-2-phenylbutanol Hydrochloride, Trimebutine Imp. A (EP) as Hydrochloride, Trimebutine Maleate Impurity A as Hydrochloride, Trimebutine Impurity A as Hydrochloride
InChI formel InChI=1S/C12H19NO.ClH/c1-4-12(10-14,13(2)3)11-8-6-5-7-9-11;/h5-9,14H,4,10H2,1-3H3;1H
LEDER Cl.CCC(CO)(N(C)C)c1ccccc1
IUPAC-namn 2-(dimethylamino)-2-phenylbutan-1-ol;hydrochloride
Molekylvikt (g/mol) 229.75
Kvantitet 25 mg
Formel vikt 229.1233 g/mol