Learn More
(2RS)-3-Bromo-2-methyl-propanoic Acid, Mikromol™
Product Code. 17952616
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity Unit Size
17952616 100 mg 100mg
1 options
Product Code. 17952616 Supplier Mikromol Supplier No. MM0067.06
 Restricted Item
Beräknad leverans: 30-11-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2RS)-3-Bromo-2-methyl-propanoic Acid
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (2RS)-3-Bromo-2-methyl-propanoic Acid
CAS 56970-78-6
Analyte or Component Names (2RS)-3-Bromo-2-methyl-propanoic Acid
Shipping Condition Room Temperature
Molecular Formula C4 H7 Br O2
Certifications/Compliance ISO 9001
Synonym (2RS)-3-Bromo-2-methyl-propanoic Acid, Captopril Imp. D (EP), Captopril Impurity D
InChI Formula InChI=1S/C4H7BrO2/c1-3(2-5)4(6)7/h3H,2H2,1H3,(H,6,7)
SMILES CC(CBr)C(=O)O
IUPAC Name 3-bromo-2-methylpropanoic acid
Molecular Weight (g/mol) 167
Quantity 100 mg
Formula Weight 165.9629 g/mol