Läs mer
(2RS)-3-(Naphthalen-1-yloxy)propane-1,2-diol (Diol Derivative), Mikromol™
Produktkod. 18023860
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18023860 25 mg
17909979 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18023860 Leverantör Mikromol Leverantörsnummer MM0043.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2RS)-3-(Naphthalen-1-yloxy)propane-1,2-diol (Diol Derivative)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (2RS)-3-(Naphthalen-1-yloxy)propane-1,2-diol
CAS 36112-95-5
Analyt- eller komponentnamn (2RS)-3-(Naphthalen-1-yloxy)propane-1,2-diol
Frakt skick Room Temperature
Molekylformel C13 H14 O3
Certifikat och efterlevnad ISO 17025
Synonym Propranolol Hydrochloride Imp. A (EP), Propranolol Imp. A (EP), (2RS)-3-(Naphthalen-1-yloxy)propane-1,2-diol, Diol Derivative of Propranolol, Propranolol Hydrochloride Impurity A, Propranolol Impurity A
InChI formel InChI=1S/C13H14O3/c14-8-11(15)9-16-13-7-3-5-10-4-1-2-6-12(10)13/h1-7,11,14-15H,8-9H2
LEDER OCC(O)COc1cccc2ccccc12
IUPAC-namn 3-naphthalen-1-yloxypropane-1,2-diol
Molekylvikt (g/mol) 218.25
Kvantitet 25 mg
Formel vikt 218.0943 g/mol