Läs mer
(2RS,4S)-5,5-Dimethyl-2-[[(phenoxyacetyl)amino]methyl]thiazolidine-4-carboxylic Acid (Penilloic Acids of Phenoxymethylpenicillin), Mikromol™
Produktkod. 30049810
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30049810 100 mg
30050015 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30049810 Leverantör Mikromol Leverantörsnummer MM0026.05
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2RS,4S)-5,5-Dimethyl-2-[[(phenoxyacetyl)amino]methyl]thiazolidine-4-carboxylic Acid (Penilloic Acids of Phenoxymethylpenicillin)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Penilloic Acids of Phenoxymethylpenicillin
CAS 2714805-53-3
Analyt- eller komponentnamn Penilloic Acids of Phenoxymethylpenicillin
Frakt skick Room Temperature
Molekylformel C15 H20 N2 O4 S
Certifikat och efterlevnad ISO 17025
Synonym Phenoxymethylpenicillin Potassium Imp. F (EP), Phenoxymethylpenicillin Imp. F (EP), (2RS,4S)-5,5-Dimethyl-2-[[(phenoxyacetyl)amino]methyl]thiazolidine-4-carboxylic Acid, Penilloic Acids of Phenoxymethylpenicillin, Phenoxymethylpenicillin Potassium Impurity F, Phenoxymethylpenicillin Impurity F
InChI formel InChI=1S/C22H28N2O2.ClH/c1-2-22(25)23(20-11-7-4-8-12-20)21-14-17-24(26,18-15-21)16-13-19-9-5-3-6-10-19;/h3-12,21H,2,13-18H2,1H3;1H
LEDER CC1(C)SC(CNC(=O)COc2ccccc2)N[C@H]1C(=O)O
IUPAC-namn (4S)-5,5-dimethyl-2-[[(2-phenoxyacetyl)amino]methyl]-1,3-thiazolidine-4-carboxylic acid
Molekylvikt (g/mol) 324.4
Kvantitet 100 mg
Formel vikt 324.1144 g/mol