Läs mer
(2RS)-N-[4-Cyano-3-(trifluoromethyl)phenyl]-3-[(4-fluorophenyl)sulfanyl]-2-hydroxy-2-methylpropanamide, Mikromol™
Produktkod. 30461527
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30461527 4 x 25 mg
30051268 25 mg
30050167 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30461527 Leverantör Mikromol Leverantörsnummer MM0894.1200254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2RS)-N-[4-Cyano-3-(trifluoromethyl)phenyl]-3-[(4-fluorophenyl)sulfanyl]-2-hydroxy-2-methylpropanamide, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (2RS)-N-[4-Cyano-3-(trifluoromethyl)phenyl]-3-[(4-fluorophenyl)sulfanyl]-2-hydroxy-2-methylpropanamide
CAS 90356-78-8
Analyt- eller komponentnamn (2RS)-N-[4-Cyano-3-(trifluoromethyl)phenyl]-3-[(4-fluorophenyl)sulfanyl]-2-hydroxy-2-methylpropanamide
Frakt skick Room Temperature
Molekylformel C18 H14 F4 N2 O2 S
Certifikat och efterlevnad ISO 17025
Synonym N-[4-Cyano-3-(trifluoromethyl)phenyl]-3-[(4-fluorophenyl)thio]-2-hydroxy-2-methylpropanamide, N-[4-Cyano-3-(trifluoromethyl)phenyl]-3-(4-fluorophenylsulfanyl)-2-hydroxy-2-methylpropionamide, Bicalutamide Imp. J (EP), Propanamide, N-[4-cyano-3-(trifluoromethyl)phenyl]-3-[(4-fluorophenyl)thio]-2-hydroxy-2-methyl-, (+/-)-, Bicalutamide Impurity J
InChI formel InChI=1S/C18H14F4N2O2S/c1-17(26,10-27-14-6-3-12(19)4-7-14)16(25)24-13-5-2-11(9-23)15(8-13)18(20,21)22/h2-8,26H,10H2,1H3,(H,24,25)
Lösningstyp Neat
LEDER CC(O)(CSc1ccc(F)cc1)C(=O)Nc2ccc(C#N)c(c2)C(F)(F)F
IUPAC-namn N-[4-cyano-3-(trifluoromethyl)phenyl]-3-(4-fluorophenyl)sulfanyl-2-hydroxy-2-methylpropanamide
Molekylvikt (g/mol) 398.37
Kvantitet 4 x 25 mg
Formel vikt 398.0712 g/mol