Läs mer
(2S)-1,2,3,4-Tetrahydro-4-oxo-2-naphthalenecarboxylic Acid, TRC
Produktkod. 30415448
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30415448 1 mg
30421472 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30415448 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT1784501MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 113867-24-6
Molekylformel C11H10O3
Molekylvikt (g/mol) 190.2
Synonym (2S)-4-Oxo-1,2,3,4-tetrahydronaphthalene-2-carboxylic acid
IUPAC-namn (2S)-4-oxo-2,3-dihydro-1H-naphthalene-2-carboxylic acid
LEDER OC(=O)[C@@H]1CC(=O)c2ccccc2C1
Specifikationer
Kemiskt namn eller material (2S)-1,2,3,4-Tetrahydro-4-oxo-2-naphthalenecarboxylic Acid
Kvantitet 1 mg
InChI formel InChI=1S/C11H10O3/c12-10-6-8(11(13)14)5-7-3-1-2-4-9(7)10/h1-4,8H,5-6H2,(H,13,14)/t8-/m0/s1
Formel vikt 190.063