missing translation for 'onlineSavingsMsg'
Learn More

(2S)-1,2,3,6-Tetrahydro-4-methyl-1-[(1R)-1-phenylethyl]-2-pyridinecarboxylic acid Ethyl Ester, TRC

Product Code. 30424057
Change view
Click to view available options
Kvantitet:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30424057 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30424057 Supplier Toronto Research Chemicals Supplier No. TRCT2940151G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material (2S)-1,2,3,6-Tetrahydro-4-methyl-1-[(1R)-1-phenylethyl]-2-pyridinecarboxylic acid Ethyl Ester
CAS 134984-61-5
Molekylformel C17H23NO2
Kvantitet 1 g
Synonym [R-(R*,S*)]-1,2,3,6-Tetrahydro-4-methyl-1-(1-phenylethyl)-2-pyridinecarboxylic Acid Ethyl Ester
InChI formel InChI=1S/C17H23NO2/c1-4-20-17(19)16-12-13(2)10-11-18(16)14(3)15-8-6-5-7-9-15/h5-10,14,16H,4,11-12H2,1-3H3/t14-,16+/m1/s1
LEDER CCOC(=O)[C@@H]1CC(=CCN1[C@H](C)c2ccccc2)C
IUPAC-namn ethyl (2S)-4-methyl-1-[(1R)-1-phenylethyl]-3,6-dihydro-2H-pyridine-2-carboxylate
Molekylvikt (g/mol) 273.37
Formel vikt 273.1729
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.