Läs mer
(2S)-2-[(3S,8aS)-3-(4-Aminobutyl)-1,4-dioxohexahydropyrrolo[1,2-a]pyrazin-2(1H)-yl]-4-phenylbutanoic Acid (S,S,S-Diketopiperazine), Mikromol™
Produktkod. 17931396
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
17931396 25 mg
1 options
Produktkod. 17931396 Leverantör Mikromol Leverantörsnummer MM0598.03
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2S)-2-[(3S,8aS)-3-(4-Aminobutyl)-1,4-dioxohexahydropyrrolo[1,2-a]pyrazin-2(1H)-yl]-4-phenylbutanoic Acid (S,S,S-Diketopiperazine)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (2S)-2-[(3S,8aS)-3-(4-Aminobutyl)-1,4-dioxohexahydropyrrolo[1,2-a]pyrazin-2(1H)-yl]-4-phenylbutanoic Acid (S,S,S-Diketopiperazine)
CAS 328385-86-0
Analyt- eller komponentnamn (2S)-2-[(3S,8aS)-3-(4-Aminobutyl)-1,4-dioxohexahydropyrrolo[1,2-a]pyrazin-2(1H)-yl]-4-phenylbutanoic Acid (S,S,S-Diketopiperazine)
Frakt skick Ice pack (-20°C)
Molekylformel C21 H29 N3 O4
Certifikat och efterlevnad ISO 9001
Synonym Lisinopril Dihydrate Imp. C (EP), Lisinopril Imp. C (EP), Lisinopril USP Related Compound A, Lisinopril USP RC A, (2S)-2-[(3S,8aS)-3-(4-Aminobutyl)-1,4-dioxohexahydropyrrolo[1,2-a]pyrazin-2(1H)-yl]-4-phenylbutanoic Acid, S,S,S-Diketopiperazine, (alphaS,3S,8aS)-3-(4-Aminobutyl)hexahydro-1,4-dioxo-alpha-(2-phenylethyl)pyrrolo[1,2-a]pyrazine-2(1H)-acetic acid, (2S)-2-[(3S,8aS)-3-(4-Aminobutyl)-1,4-dioxohexahydropyrrolo[1,2-a]pyrazin-2(H)-yl]-4-phenylbutanoic acid, Lisinopril Related Compound A, Lisinopril Dihydrate Impurity C, Lisinopril Impurity C
InChI formel InChI=1S/C21H29N3O4/c22-13-5-4-9-17-19(25)23-14-6-10-16(23)20(26)24(17)18(21(27)28)12-11-15-7-2-1-3-8-15/h1-3,7-8,16-18H,4-6,9-14,22H2,(H,27,28)/t16-,17-,18-/m0/s1
LEDER NCCCC[C@@H]1N([C@@H](CCc2ccccc2)C(=O)O)C(=O)[C@@H]3CCCN3C1=O
IUPAC-namn (2S)-2-[(3S,8aS)-3-(4-aminobutyl)-1,4-dioxo-6,7,8,8a-tetrahydro-3H-pyrrolo[1,2-a]pyrazin-2-yl]-4-phenylbutanoic acid
Molekylvikt (g/mol) 387.47
Kvantitet 25 mg
Formel vikt 387.2158 g/mol