Läs mer
(2S)-2-Amino-benzenebutanoic Acid Benzyl Ester Tosylate Salt (>90%), TRC
Produktkod. 30318299
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30318299 250 mg
1 options
Produktkod. 30318299 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA591420250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (2S)-2-Amino-benzenebutanoic Acid Benzyl Ester, Tosylate Sal
CAS 117560-24-4
Molekylformel C17 H19 N O2 . C7 H8 O3 S
Kvantitet 250 mg
Synonym (2S)-2-Amino-Benzenebutanoic Acid Benzyl Ester Tosylate Salt
InChI formel InChI=1S/C17H19NO2.C7H8O3S/c18-16(12-11-14-7-3-1-4-8-14)17(19)20-13-15-9-5-2-6-10-15;1-6-2-4-7(5-3-6)11(8,9)10/h1-10,16H,11-13,18H2;2-5H,1H3,(H,8,9,10)/t16-;/m0./s1
LEDER Cc1ccc(cc1)S(=O)(=O)O.N[C@@H](CCc2ccccc2)C(=O)OCc3ccccc3
IUPAC-namn benzyl (2S)-2-amino-4-phenylbutanoate;4-methylbenzenesulfonic acid
Molekylvikt (g/mol) 441.54
Formel vikt 441.161