Läs mer
(2S)-(2-Chlorophenyl)[6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl]acetic Acid Hydrochloride (Clopidogrel Acid Hydrochloride), Mikromol™
Beskrivning
(2S)-(2-Chlorophenyl)[6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl]acetic Acid Hydrochloride (Clopidogrel Acid Hydrochloride)
Specifikationer
Specifikationer
| Kemiskt namn eller material | Clopidogrel carboxylic acid hydrochloride |
| CAS | 144750-42-5 |
| Analyt- eller komponentnamn | Clopidogrel carboxylic acid hydrochloride |
| Frakt skick | Room Temperature |
| Molekylformel | C15 H14 Cl N O2 S . Cl H |
| Certifikat och efterlevnad | ISO 17025 |
| Synonym | Clopidogrel Hydrogen Sulfate Imp. A (EP), Clopidogrel Imp. A (EP), Clopidogrel USP Related Compound A, Clopidogrel USP RC A, Thieno[3,2-c]pyridine-5(4H)-acetic acid, alpha-(2-chlorophenyl)-6,7-dihydro-, hydrochloride (1:1), (alphaS)-, Thieno[3,2-c]pyridine-5(4H)-acetic acid, alpha-(2-chlorophenyl)-6,7-dihydro-, hydrochloride, (alphaS)- (9CI), (+)-(S)-(o-chlorophenyl)-6,7-dihydrothieno[3,2-c]pyridine-5(4H)-acetic acid, hydrochloride, Clopidogrel Related Compound A as Hydrochloride, Clopidogrel Hydrogen Sulfate Impurity A as Hydrochloride, Clopidogrel Impurity A as Hydrochloride |
| InChI formel | InChI=1S/C15H14ClNO2S.ClH/c16-12-4-2-1-3-11(12)14(15(18)19)17-7-5-13-10(9-17)6-8-20-13;/h1-4,6,8,14H,5,7,9H2,(H,18,19);1H/t14-;/m0./s1 |
| LEDER | Cl.OC(=O)[C@@H](N1CCc2sccc2C1)c3ccccc3Cl |
| IUPAC-namn | (2S)-2-(2-chlorophenyl)-2-(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)acetic acid;hydrochloride |
| Visa mer |