Läs mer
(2S)-2-Hydroxyglutaric Acid Octyl Ester Sodium Salt, TRC
Produktkod. 30008168
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008168 10 mg
30003696 5 mg
30007829 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30008168 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH94259610MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1391067-96-1
Molekylformel C13H23NaO5
Molekylvikt (g/mol) 282.31
Synonym (2S)-2-Hydroxypentanedioic Acid Octyl Ester Sodium Salt, (S)-2-Hydroxyglutaric Acid Octyl Ester Sodium Salt, (S)-α-Hydroxyglutaric Acid Octyl Ester Sodium Salt, L-2-Hydroxyglutaric Acid Octyl Ester Sodium Salt, Octyl L-2-Hydroxyglutarate Sodium Salt, Octyl-L-2-HG Sodium;
IUPAC-namn sodium;(4S)-4-hydroxy-5-octoxy-5-oxopentanoate
LEDER CCCCCCCCOC(=O)[C@H](CCC(=O)[O-])O.[Na+]
Specifikationer
Kemiskt namn eller material (2S)-2-Hydroxyglutaric Acid Octyl Ester Sodium Salt
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C13H24O5.Na/c1-2-3-4-5-6-7-10-18-13(17)11(14)8-9-12(15)16;/h11,14H,2-10H2,1H3,(H,15,16);/q;+1/p-1/t11-;/m0./s1
Formel vikt 282.1443
Procent renhet >95%