Läs mer
(2S)-3-Azido-2-hydroxy-2-methyl-propanoic Acid Methyl Ester, TRC
Produktkod. 30330065
Change view
Kvantitet:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30330065 250 mg
30345628 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30330065 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA850110250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (2S)-3-Azido-2-hydroxy-2-methyl-propanoic Acid Methyl Ester
CAS 549504-45-2
Molekylformel C5 H9 N3 O3
Kvantitet 250 mg
InChI formel InChI=1S/C5H9N3O3/c1-5(10,3-7-8-6)4(9)11-2/h10H,3H2,1-2H3/t5-/m0/s1
LEDER COC(=O)[C@@](C)(O)CN=[N+]=[N-]
IUPAC-namn methyl (2S)-3-azido-2-hydroxy-2-methylpropanoate
Molekylvikt (g/mol) 159.14
Formel vikt 159.0644